mono-tert-Butyl malonate, 95%, Thermo Scientific Chemicals
mono-tert-Butyl malonate, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

mono-tert-Butyl malonate, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
10 g
Packaging:
Glass bottle
Catalog number 438830010
Price (USD)/ Each
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Name3-(tert-butoxy)-3-oxopropanoic acid
Molecular FormulaC7H12O4
InChI KeyNGGGZUAEOKRHMA-UHFFFAOYSA-N
SMILESCC(C)(C)OC(=O)CC(O)=O
Molecular Weight (g/mol)160.17
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Infrared spectrumConforms
Appearance (Form)Liquid or low melting solid
Titration with NaOH94.0 to 106.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only