Thermo Scientific Chemicals

1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide, 97%, Thermo Scientific Chemicals

Catalog number: 438842500
250 g, Glass bottle, Each
Thermo Scientific Chemicals

1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide, 97%, Thermo Scientific Chemicals

Catalog number: 438842500
250 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 438842500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1892-57-5
IUPAC Name
({[3-(dimethylamino)propyl]imino}methylidene)(ethyl)amine
Molecular Formula
C8H17N3
InChI Key
LMDZBCPBFSXMTL-UHFFFAOYSA-N
SMILES
CCN=C=NCCCN(C)C
Appearance (Color)
Colorless to light yellow
Infrared spectrum
Conforms
GC
>=96.0 %
Appearance (Form)
Liquid
Refractive index
1.4600 to 1.4630 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Coupling reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text