Thermo Scientific Chemicals

(1,3-Dioxolan-2-ylmethyl)magnesium bromide, 0.5M solution in THF, AcroSeal™, Thermo Scientific Chemicals

Catalog number: 438861000
100 mL, AcroSeal™ Glass Bottle, Each
Thermo Scientific Chemicals

(1,3-Dioxolan-2-ylmethyl)magnesium bromide, 0.5M solution in THF, AcroSeal™, Thermo Scientific Chemicals

Catalog number: 438861000
100 mL, AcroSeal™ Glass Bottle, Each
Quantity
Packaging
Catalog number: 438861000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
180675-22-3
IUPAC Name
2-[(bromomagnesio)methyl]-1,3-dioxolane
Molecular Formula
C4H7BrMgO2
InChI Key
ZDNTZRAFJZERSL-UHFFFAOYSA-M
SMILES
Br[Mg]CC1OCCO1
Appearance (Color)
Clear to turbid yellow to green to brown
Appearance
May develop some turbidity or precipitate
Appearance (Form)
Solution
Titration Grignard
0.40 to 0.60 M

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text