Specifications
For Use With (Equipment)7500 Fast System, 7500 System, 7900HT Fast System, 7900HT System,Applied Biosystems StepOnePlus™ Fast Real-Time PCR System, StepOne™, Standard Mode, StepOnePlus™, Standard Mode
FormatTube
Hot StartBuilt-In Hot Start
No. of Reactions500 Reactions
Passive Reference DyeROX (Pre-mixed)
PolymeraseAmpliTaq Gold DNA Polymerase
Product LineAmbion™, Path-ID™
Purity or Quality GradeUP (Ultra Pure)
Quantity500 reactions
Sample TypeDNA (Genomic), cDNA
Shipping ConditionDry Ice
Detection MethodPrimer-probe
For Use With (Application)Animal Health, Pathogen Detection
FormLiquid
PCR MethodqPCR
Reaction SpeedFast or Standard
Unit SizeEach