Thermo Scientific Chemicals

Cyclopropanesulfonyl chloride, 97%, Thermo Scientific Chemicals

Catalog number: 439020050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

Cyclopropanesulfonyl chloride, 97%, Thermo Scientific Chemicals

Catalog number: 439020050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 439020050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
139631-62-2
IUPAC Name
cyclopropanesulfonyl chloride
Molecular Formula
C3H5ClO2S
InChI Key
PFWWSGFPICCWGU-UHFFFAOYSA-N
SMILES
ClS(=O)(=O)C1CC1
Appearance (Form)
Liquid
Infrared spectrum
Conforms
Appearance (Color)
Clear colorless to light yellow
GC
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text