Thermo Scientific Chemicals

BES sodium salt, 99%

Catalog number: 439035000
500 g, Each
Thermo Scientific Chemicals

BES sodium salt, 99%

Catalog number: 439035000
500 g, Each
Quantity
Catalog number: 439035000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
66992-27-6
IUPAC Name
sodium 2-[bis(2-hydroxyethyl)amino]ethane-1-sulfonate
Molecular Formula
C6H14NNaO5S
InChI Key
CFQLQLSIZOWFNV-UHFFFAOYSA-M
SMILES
[Na+].OCCN(CCO)CCS([O-])(=O)=O
Appearance (Color)
White
Elemental Analysis
5.85 to 6.04 % (N)
Appearance (Form)
Powder or solid
Infrared spectrum
Conforms
Titration with HCl
98.5 to 101.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological buffer
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text