Informational:Inquire about OEM or Commercial Supply version of this product here.
Informational:Upgrade to Platinum II Taq Hot-Start DNA Polymerase for universal annealing and a 4X faster cycling protocol.
Applied Biosystems™

AmpliTaq Gold™ Fast PCR Master Mix

Catalog number: 4390939
Have Questions?
Applied Biosystems™

AmpliTaq Gold™ Fast PCR Master Mix

Catalog number: 4390939
Have Questions?
Catalog Number
4390939
Unit Size
250 reactions
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
25 reactions
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
2,500 reactions
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
4390939250 reactions
-
439093725 reactions
-
43909412,500 reactions
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The Applied Biosystems™ AmpliTaq Gold™ Fast PCR Master Mix is a premix, hot start chemistry for end-point PCR analysis. Its optimized chemical design enables to shorten PCR running time from a traditional 2 hours to proximal 40 minutes. It generates high quality PCR products that give high quality sequencing results that customers expect from Applied Biosystems™. Optimized for use in sequencing applications, this kit provides specific, high-yield amplicons that can be easily and robustly sequenced. This premixed master mix works with your existing primers to provide sensitive, specific and reproducible results.

• Fast Results: Speeds the time from sample to results. Amplifies 600bp length gene in approximately 45 minutes

• Better Sequencing quality: Specific, high-yield amplicons suitable for sequencing applications that yield low peak-under-peak, longer continous read-length and higher QV-30 count

• Easy to use: The 2X master mix allows for quick experiment setup

• No primer annealing restrictions

Speed Time to Discovery
AmpliTaq Gold™ Fast PCR Master Mix, UP brings PCR reaction times down to as little as 45 minutes and delivers robust sequencing data on par with standard PCR reagents such as the classic AmpliTaq Gold™ PCR Master Mix. This new master mix has been designed for use with your existing primers and results in specific, high-yield amplicons for easy, high quality sequencing data. Cycle sequencing can now also be performed in as little as 50 minutes.

Quick Setup, High Sensitivity
This pre-mixed hotstart kit allows you to setup your experiment quickly without sacrificing performance. This kit provides reproducible, sensitive results, allowing you to detect a single copy of a gene in 10 ng of DNA.

Robust Fast Sequencing Results
Get the sequencing results you expect from Applied Biosystems™. The AmpliTaq Gold™ Fast PCR Master Mix, UP is optimized for your sequencing reactions. This kit yields specific high-yield amplicons that provide you with clean sequencing reads including low peak-under-peak, longer read-lengths and superior Q30.

Note: See user's manual or package insert for limited label license, and trademark information. For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Detection Method
Primer-probe
Format
Tube
GC-Rich PCR Performance
High
Polymerase
AmpliTaq Gold DNA Polymerase
Reaction Speed
Fast
Exonuclease Activity
5' - 3'
Product Line
AmpliTaq Gold™
Product Type
Fast PCR Master Mix
Quantity
250 Reactions
Shipping Condition
Wet Ice
Sufficient For
250 Reactions
For Use With (Application)
Cloning
Concentration
2X
Fidelity (vs. Taq)
1 X
Hot Start
Built-In Hot Start
No. of Reactions
250 Reactions
Overhang
3'-A
Reaction Format
SuperMix or Master Mix
Size (Final Product)
1.5 kb or less
Starting Material
DNA
Volume
2.5 mL

Contents & Storage

This master mix comes in 2 x 1.25 mL tubes, enough for 250 reactions.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text