Ethynylmagnesium bromide, 0.5M solution in THF, AcroSeal™, Thermo Scientific Chemicals
Ethynylmagnesium bromide, 0.5M solution in THF, AcroSeal™, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ethynylmagnesium bromide, 0.5M solution in THF, AcroSeal™, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle
Catalog number 439121000
Price (USD)/ Each
54.65
Online exclusive
64.70 
Save 10.05 (16%)
-
Add to cart
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (USD)/ Each
54.65
Online exclusive
64.70 
Save 10.05 (16%)
Add to cart
Ethynylmagnesium bromide, 0.5M solution in THF, AcroSeal™, Thermo Scientific Chemicals
Catalog number439121000
Price (USD)/ Each
54.65
Online exclusive
64.70 
Save 10.05 (16%)
-
Add to cart
Chemical Identifiers
IUPAC Namebromo(ethynyl)magnesium
Molecular FormulaC2HBrMg
InChI KeyHUGJUYPSXULVQQ-UHFFFAOYSA-M
SMILESBr[Mg]C#C
Molecular Weight (g/mol)129.24
View more
SpecificationsSpecification SheetSpecification Sheet
Titration Grignard0.40 to 0.60 M
AppearanceIf precipitation is present, gently mix
Appearance (Form)Solution
Appearanceprecipitate back into solution under an inert
Appearanceatmosphere
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only