Thermo Scientific Chemicals

Magnesium carbonate, for biochemistry, specified according to the requirements of US, Thermo Scientific Chemicals

Catalog number: 439130010
1 kg, Plastic bottle, Each
Thermo Scientific Chemicals

Magnesium carbonate, for biochemistry, specified according to the requirements of US, Thermo Scientific Chemicals

Catalog number: 439130010
1 kg, Plastic bottle, Each
Quantity
Packaging
Catalog number: 439130010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
546-93-0
IUPAC Name
magnesium(2+) carbonate
Molecular Formula
CMgO3
InChI Key
ZLNQQNXFFQJAID-UHFFFAOYSA-L
SMILES
[Mg++].[O-]C([O-])=O
Appearance (Color)
White
Appearance (Form)
Powder
Additional info
Absence-specified microorganisms: Passes
Calcium (Ca)
=<0.45 %
Iron (Fe)
=<0.02 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For biochemistry
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text