Chemical Identifiers
CAS1787-61-7
IUPAC Namesodium (4Z)-4-[2-(1-hydroxynaphthalen-2-yl)hydrazin-1-ylidene]-7-nitro-3-oxo-3,4-dihydronaphthalene-1-sulfonate
Molecular FormulaC20H12N3NaO7S
InChI KeyJHUJLRKQZAPSDP-GXTSIBQPSA-M
SMILES[Na+].OC1=C2C=CC=CC2=CC=C1N\N=C1/C(=O)C=C(C2=CC(=CC=C12)[N+]([O-])=O)S([O-])(=O)=O
View more