Chemical Identifiers
CAS811862-77-8
IUPAC Namebis(1,5-bis(3,5-dimethoxyphenyl)penta-1,4-dien-3-one) palladium
Molecular FormulaC42H44O10Pd
InChI KeyRTGAJOPJZJDWAX-UHFFFAOYSA-N
SMILES[Pd].COC1=CC(C=CC(=O)C=CC2=CC(OC)=CC(OC)=C2)=CC(OC)=C1.COC1=CC(C=CC(=O)C=CC2=CC(OC)=CC(OC)=C2)=CC(OC)=C1
View more