Informational:Inquire about OEM or Commercial Supply version of this product here.
Invitrogen™

TaqMan™ MicroRNA Cells-to-CT™ Kit

Catalog number: 4391848
Have Questions?
Invitrogen™

TaqMan™ MicroRNA Cells-to-CT™ Kit

Catalog number: 4391848
Have Questions?
Catalog Number
4391848
Unit Size
100 reactions
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
4391848100 reactions
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The Ambion™ TaqMan™ MicroRNA Cells-to-CT™ Kit makes it possible to perform microRNA (miRNA) expression profiling directly from cultured cells without RNA purification. The kit is the first to offer a simple, complete workflow for miRNA expression analysis from a few samples, or it can easily be incorporated into automated, high throughput applications.

• Complete, validated solution—Optimized workflow includes cell lysis reagents, DNase, TaqMan™ MicroRNA RT reagents, and TaqMan™ Universal PCR Master Mix
• Fast, simple, and convenient—Prepare samples in 10 minutes; eliminate tedious RNA isolation
• Gold standard specificity—Achieve superior results with TaqMan™ MicroRNA reagents
• Robust performance—Results equivalent to purified RNA
• Broad input dynamic range—Linear detection from 10 to 100,000 cells

Complete, Validated Solution

Featuring a unique method for lysing cultured cells while removing genomic DNA and preserving RNA integrity, the TaqMan™ MicroRNA Cells-to-CT™ Kit contains TaqMan™ MicroRNA reverse transcription (RT) reagents and TaqMan™ Universal PCR Master Mix for real-time PCR analysis. The TaqMan™ MicroRNA Cells-to-CT™ Kit has been extensively tested and validated with TaqMan™ MicroRNA Assays, sold separately.

Simple, Convenient 10-Minute Sample Preparation

The TaqMan™ MicroRNA Cells-to-CT™ Kit incorporates a simple 10-minute sample preparation procedure (Figure 1) for use with 10–100,000 cultured cells⁄sample, either in plates or tubes. Cells are washed in PBS and lysed for 8 minutes at room temperature; DNase treatment can be performed concurrently. Lysis is terminated at room temperature by a 2-minute incubation with Stop Solution.

The TaqMan™ MicroRNA Cells-to-CT™ Kit lysates are now ready for reverse transcription or can be stored at -20°C for up to 5 months. Since samples are not subjected to an RNA purification process, all of the small RNA species are available for analysis. Because samples can be processed directly in culture plates (96 or 384 wells), sample handling is reduced, and the risk of sample loss or transfer error is minimized. No heating, washing, or centrifugation is required; the kit greatly simplifies a traditionally time-consuming, labor intensive process and reduces it to 10 minutes.

Gold Standard Specificity—Superior Results with TaqMan™ MicroRNA Reagents

Applied Biosystems has developed a state-of-the-art solution to the fundamental problem associated with the short length of mature miRNAs. TaqMan™ MicroRNA Assays (sold separately) include a unique, target-specific stem-loop reverse-transcription primer (Figure 2). The stem-loop structure is designed to incorporate specificity for the targeted mature miRNA, in contrast to other methods that indiscriminately extend and prime all RNA in a sample. The TaqMan™ MicroRNA RT reagents, included in the TaqMan™ MicroRNA Cells-to-CT™ Kit, were specifically designed to work in conjunction with this innovative assay design.

The ability to distinguish between highly homologous mature miRNA targets is required for accurate miRNA expression analysis. The TaqMan™ MicroRNA Cells-to-CT™ Kit, in conjunction with the TaqMan™ MicroRNA Assays, delivers the specificity needed to distinguish between highly homologous targets, as seen in Figure 3. In this example, lysates from the TaqMan™ MicroRNA Cells-to-CT™ Kit or purified miRNA were used in real-time PCR using TaqMan™ MicroRNA Assays to detect single base differences on exogenously added miRNA templates. Specificity obtained with TaqMan™ MicroRNA Cells-to-CT™ Kit lysates (Figure 3A) was equivalent to that using purified RNA (Figure 3B).

Robust Performance—Results Equivalent to Purified RNA

This new kit has been extensively tested with a broad selection of TaqMan™ MicroRNA Assays and demonstrates performance equivalent to that obtained with purified RNA (Figure 4). The TaqMan™ MicroRNA Cells-to-CT™ Kit workflow enables unsurpassed miRNA expression evaluation with > 900 available TaqMan™ MicroRNA Assays.

Broad Input Dynamic Range—Linear Detection from 10 to 100,000 Cells

Because miRNA expression levels can vary widely due to sample type, developmental stage, or disease progression, it is important to have sensitive tools for miRNA expression analysis. Now, miRNA targets that vary from a few copies to millions of copies can be accurately quantitated in the same experiment.

The TaqMan™ MicroRNA Cells-to-CT™ Kit works robustly with as few as 10 cells, and up to 100,000 cells. Furthermore, to maximize sensitivity, the TaqMan™ MicroRNA Cells-to-CT™ Kit allows up to 33% of the RT reaction volume to be comprised of sample lysate (singleplex RT). Figure 5 demonstrates the linearity and sensitivity of detection from TaqMan™ MicroRNA Cells-to-CT™ Kit lysates across 5 logs of cellular input per lysis reaction; results were equivalent to purified RNA. Additionally, the broad dynamic range of TaqMan™ MicroRNA Assays is not compromised by TaqMan™ MicroRNA Cells-to-CT™ Kit lysates (data not shown).

Proven Performance, Proven Together

All components of the TaqMan™ MicroRNA Cells-to-CT™ Kit have been optimized for consistent and reliable performance. This removes the guesswork involved in assembling separate kits for sample preparation, reverse transcription, and real-time PCR. The TaqMan™ MicroRNA Cells-to-CT™ Kit has been validated with TaqMan™ MicroRNA Assays for added quality assurance.
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Detection Method
Primer-probe
Format
Tube
GC-Rich PCR Performance
High
PCR Method
2-step RT-qPCR
Polymerase
DNA Polymerase
Reaction Speed
Standard
For Use With (Equipment)
7000 System, 7300 System, 7500 System, 7900HT System,Applied Biosystems StepOnePlus™ Fast Real-Time PCR System, StepOne™, Standard Mode, StepOnePlus™, Standard Mode, QuantStudio 6 and 7 Pro systems, QuantStudio 7 Flex system, QuantStudio 3 and 5 systems, QuantStudio 12k Flex system, QuantStudio 6 Flex system
Green Features
Fewer resources used and less waste, Less hazardous
Passive Reference Dye
ROX (Pre-mixed)
Product Line
Ambion™, Cells-to-CT™, TaqMan™
Product Type
MicroRNA Cells-to-C Kit
Reverse Transcriptase
M-MLV
Shipping Condition
Dry Ice
Sufficient For
100 Reactions
For Use With (Application)
miRNA analysis
No. of Reactions
100 Reactions
Sample Type
Cell

Contents & Storage

Includes cell lysis reagents, DNase, TaqMan™ MicroRNA RT reagents, and TaqMan™ Universal PCR Master Mix.

• 100 sample prep reactions
• 100 DNase reactions
• 100 reverse transcription reactions,
• 500 x 20 μL PCR reactions

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text