Informational:Inquire about OEM or Commercial Supply version of this product here.
Invitrogen™

Cells-to-CT Bulk RT Reagents

Catalog number: 4391852C
Have Questions?
Invitrogen™

Cells-to-CT Bulk RT Reagents

Catalog number: 4391852C
Have Questions?
Catalog Number
4391852C
Unit Size
2,500 reactions
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
4391852C2,500 reactions
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Cells-to-CT Bulk RT Reagents are separately-available, large-quantity versions of the reagents used for reverse transcription in the TaqMan™ Gene Expression Cells-to-CT™ Kit (20X RT enzyme mix and 20X RT buffer). These reagents are sufficient for 2500 RT reactions. When combined with the Cells-to- CT Bulk Lysis Reagents, these reagents can be easily incorporated into automated, high throughput applications.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Form
Liquid
Green Features
Fewer resources used and less waste, Less hazardous
Product Line
Cells-to-CT™
Product Type
cDNA Synthesis Reagents
Shipping Condition
Dry Ice
Concentration
20X
No. of Reactions
2500 reactions

Contents & Storage

• 20X RT Enzyme Mix, Store at -20°C
• 2X RT Buffer, Store at 4°C

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text