Chemical Identifiers
IUPAC Namemethyl (2S)-2-{[(tert-butoxy)carbonyl]amino}-2-(dimethoxyphosphoryl)acetate
Molecular FormulaC10H20NO7P
InChI KeyLJHAPRKTPAREGO-ZETCQYMHSA-N
SMILESCOC(=O)[C@@H](NC(=O)OC(C)(C)C)P(=O)(OC)OC
Molecular Weight (g/mol)297.24
View more