L-Lactic acid sodium salt, 99%, extra pure, Thermo Scientific Chemicals
L-Lactic acid sodium salt, 99%, extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals

L-Lactic acid sodium salt, 99%, extra pure, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Packaging:
Glass bottle
Catalog number 439220100
Price (USD)/ Each
99.65
Online exclusive
111.00 
Save 11.35 (10%)
-
Add to cart
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
99.65
Online exclusive
111.00 
Save 11.35 (10%)
Add to cart
L-Lactic acid sodium salt, 99%, extra pure, Thermo Scientific Chemicals
Catalog number439220100
Price (USD)/ Each
99.65
Online exclusive
111.00 
Save 11.35 (10%)
-
Add to cart
Chemical Identifiers
CAS867-56-1
IUPAC Namesodium 2-hydroxypropanoate
Molecular FormulaC3H5NaO3
InChI KeyNGSFWBMYFKHRBD-UHFFFAOYNA-M
SMILES[Na+].CC(O)C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point160°C to 166°C
Appearance (Color)White to light yellow
Water=<1.0 %
Titration with HClO4>=98.5 %
Specific optical rotation-12° to -13° (20°C, 589 nm) (C=1 in H2O)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only