Thermo Scientific Chemicals

2-Iodo-5-methylthiophene, 97%, Thermo Scientific Chemicals

Catalog number: 439270010
1 g, Each
Thermo Scientific Chemicals

2-Iodo-5-methylthiophene, 97%, Thermo Scientific Chemicals

Catalog number: 439270010
1 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: 439270010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
16494-36-3
IUPAC Name
2-iodo-5-methylthiophene
Molecular Formula
C5H5IS
InChI Key
NAZNQEXKAPLVKC-UHFFFAOYSA-N
SMILES
CC1=CC=C(I)S1
Appearance (Color)
Yellow to orange-red
Infrared spectrum
Conforms
Appearance (Form)
Liquid
GC
>=96.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text