Specifications
For Use With (Equipment)7000 System, 7300 System, 7500 System, 7700 System, 7900HT System,Applied Biosystems StepOnePlus™ Fast Real-Time PCR System, StepOne™, Standard Mode, StepOnePlus™, Standard Mode
No. of Reactions200 Reactions
Passive Reference DyeROX (Pre-mixed)
PolymeraseAmpliTaq Gold DNA Polymerase
Product LineRNA-to-CT™, TaqMan™
Product TypeOne-Step RT-PCR Kit
Purity or Quality GradeUP (Ultra Pure)
Quantity200 reactions
Reverse TranscriptaseArrayScript™ UP
Sample TypeRNA
Shipping ConditionWet Ice
Sufficient For200 Reactions
Detection MethodPrimer-probe
For Use With (Application)Gene Expression
PCR Method1-step RT-qPCR
Reaction SpeedStandard
Unit SizeEach