Thermo Scientific Chemicals

(+)-Biotin N-hydroxysuccinimide ester, 98%

Catalog number: 439300010
1 g, Glass bottle, Each
Thermo Scientific Chemicals

(+)-Biotin N-hydroxysuccinimide ester, 98%

Catalog number: 439300010
1 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 439300010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
35013-72-0
IUPAC Name
2,5-dioxopyrrolidin-1-yl 5-{2-oxo-hexahydro-1H-thieno[3,4-d]imidazol-4-yl}pentanoate
Molecular Formula
C14H19N3O5S
InChI Key
YMXHPSHLTSZXKH-UHFFFAOYNA-N
SMILES
O=C(CCCCC1SCC2NC(=O)NC12)ON1C(=O)CCC1=O
Appearance (Form)
Powder
Elemental Analysis
12.00 to 12.62 % N
Infrared spectrum
Conforms
Elemental Analysis
48.03 to 50.49 % C
Appearance (Color)
White to off-white

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text