(+)-Biotin N-hydroxysuccinimide ester, 98%
(+)-Biotin N-hydroxysuccinimide ester, 98%
Thermo Scientific Chemicals

(+)-Biotin N-hydroxysuccinimide ester, 98%

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 439302500
Price (USD)/ Each
224.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
264.00 
Save 39.35 (15%)
-
Add to cart
Quantity:
250 mg
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
224.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
264.00 
Save 39.35 (15%)
Add to cart
(+)-Biotin N-hydroxysuccinimide ester, 98%
Catalog number439302500
Price (USD)/ Each
224.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
264.00 
Save 39.35 (15%)
-
Add to cart
Chemical Identifiers
CAS35013-72-0
IUPAC Name2,5-dioxopyrrolidin-1-yl 5-{2-oxo-hexahydro-1H-thieno[3,4-d]imidazol-4-yl}pentanoate
Molecular FormulaC14H19N3O5S
InChI KeyYMXHPSHLTSZXKH-UHFFFAOYNA-N
SMILESO=C(CCCCC1SCC2NC(=O)NC12)ON1C(=O)CCC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Elemental Analysis12.00 to 12.62 % N
Infrared spectrumConforms
Elemental Analysis48.03 to 50.49 % C
Appearance (Color)White to off-white
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only