(S)-(+)-1-Methyl-3-pyrrolidinol, 98%, ee 99%, Thermo Scientific Chemicals
(S)-(+)-1-Methyl-3-pyrrolidinol, 98%, ee 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

(S)-(+)-1-Methyl-3-pyrrolidinol, 98%, ee 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 439320050
Price (USD)/ Each
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
IUPAC Name1-methylpyrrolidin-3-ol
Molecular FormulaC5H11NO
InChI KeyFLVFPAIGVBQGET-UHFFFAOYNA-N
SMILESCN1CCC(O)C1
Molecular Weight (g/mol)101.15
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=97.5 %
Water=<2.00 %
Appearance (Color)Colorless to yellow to brown
AppearanceMay darken on storage
Appearance (Form)Liquid
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only