Thermo Scientific Chemicals

α-Ketoglutaric acid sodium salt, 98%, Thermo Scientific Chemicals

Catalog number: 439351000
100 g, Each
Thermo Scientific Chemicals

α-Ketoglutaric acid sodium salt, 98%, Thermo Scientific Chemicals

Catalog number: 439351000
100 g, Each
Quantity
Catalog number: 439351000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
22202-68-2
IUPAC Name
disodium 2-oxopentanedioate
Molecular Formula
C5H4Na2O5
InChI Key
YBGBJYVHJTVUSL-UHFFFAOYSA-L
SMILES
[Na+].[Na+].[O-]C(=O)CCC(=O)C([O-])=O
Appearance (Color)
White
Infrared spectrum
Conforms
Loss on drying
=<1.0 %
UV
at 430 nm A: =<0.200
Titration with HClO4
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text