VeritiLink™ Remote Management Software
Product Image
Applied Biosystems™
VeritiLink™ Remote Management Software
VeritiLink™ Remote Management Software (VRMS) is an easy-to-install software that provides secure, remote management for Veriti™ Thermal Cyclers from anywhereRead more
Have Questions?
Catalog NumberQuantity
43935651 software
Catalog number 4393565
Price (USD)
5,100.00
Each
Add to cart
Quantity:
1 software
Price (USD)
5,100.00
Each
Add to cart
Ask our AI about this Product
VeritiLink™ Remote Management Software (VRMS) is an easy-to-install software that provides secure, remote management for Veriti™ Thermal Cyclers from anywhere on the network.
This easy-to-install software provides secure, remote management for Veriti™ Thermal Cyclers from anywhere on the network.
• Perform and monitor runs for up to 50+ networked Veriti™ Thermal Cyclers
• Manage all of your Veriti™ systems in a secure client-server environment with user password protection
• Receive real-time e-mail system updates on your desktop, laptop or PDA
• VeritiLink™ Remote Management Software (VRMS) browser-enabled interface means you can “Bookmark Your Veritis” for fast, convenient access
• Create and edit methods on your computer for increased security and peace of mind, and download⁄upload to⁄from your Veriti systems as needed
• Easy setup: install software on a server for use on any networked computers
Veriti Systems Anywhere, Everywhere: VeritiLink™ Remote Monitoring Software
VRMS allows you to access Veriti™ Systems anywhere, everywhere. This software package allows you to manage and monitor thermal cycling runs wherever you can access an internet browser. With this program, you can start or stop runs, create and edit methods and send them to the Veriti™ Thermal Cycler systems as needed. Designed to manage from one to 50+ Veriti™ Thermal Cyclers, the user interface shows which systems are available for use and status of systems in use to facilitate instrument sharing for busy labs. With system update email notifications to your desktop, laptop, or PDA and user password protection to keep your methods secure, this software provides you with peace of mind that your systems are running smoothly and securely.
Easy to Install
This software is easy to install, requiring only server-side installation to provide access for all networked computers. This provides more convenient access than competitors’ software which requires local computer software installation for the server computer as well as each client computer.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)PCR
For Use With (Equipment)Veriti Thermal Cycler
Product LineVeritiLink™
Quantity1 software
Software TypeRemote Management Software
Product TypeThermal Cycler Accessories
Unit SizeEach
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link