3500 Dx Series POP-7™ and POP-6™ are separation matrices for the 3500 Dx⁄3500xL Dx Genetic Analyzers. They are conveniently offered in easy-to-use pouch packages with either 960 samples (maximum) or 384 samples (maximum) size formats. Each polymer is optimized for specific applications.
Easy to Use Polymer is pre-formulated in a single use pouch, which not only saves time but also ensures reliability and consistency.
Radio Frequency Identification (RFID) label The RFID encodes easy-to-track important information such as part⁄lot number, samples remaining and expiry dates (displayed on software dashboard) Robust Formulation Polymers are formulated for a wide range of applications.
Reproducible Results POP™ polymers dynamically coat the capillary wall to control electro-osmotic flow. Their definite quality and uniform consistency eliminate guesswork and help to ensure reproducibility. POP Polymers are specifically formulated to separate DNA fragments of a known size range at a desired resolution and run time.
Many Choices POP-7™ polymer is very versatile and designed for a wide range of short-read to long-read sequencing applications. POP-6™ polymer is the choice for certain sequencing applications.
Performance Optimized Polymer This product is intended for in-vitro diagnostic use on the Applied Biosystems 3500 Dx Series Genetic Analyzers and is not validated for use with other systems or applications. The 3500 Dx Series Genetic Analyzer is to be used by trained operators in the analysis of human DNA or RNA, for the detection of genetic changes that may lead to disease presence or susceptibility. Off-label uses such as fragment analysis are for research use only. Refer to the Applied Biosystems 3500 Dx⁄3500xL Dx Genetic Analyzer User Guide for further details.
Specifications
Recommended Storage
Store the polymer pouch at 2-8°C until ready to use. Note that expired pouches cannot be used on the instrument.
On-instrument lifeA ⁄UsageB On 3500 Dx (8 capillary) Genetic Analyzer, -Lower of 7 days or 384 samples or 60 Injections On 3500xl Dx (24 capillary) Genetic Analyzer -Lower of 7 days or 384 samples or 20 Injections
The pouch includes additional polymer volume to accommodate both installation and a reasonable number of post-installation wizard operations such as bubble removal without affecting the number of samples⁄injections available.
For Use With (Application)
Long-Read Sequencing, Standard Sequencing, Short Read Sequencing, Fragment Analysis (Denaturing)