Applied Biosystems™

POP-6™ Polymer for 3500 Dx/3500xL Dx Genetic Analyzers

Catalog number: 4393711
Have Questions?
Applied Biosystems™

POP-6™ Polymer for 3500 Dx/3500xL Dx Genetic Analyzers

Catalog number: 4393711
Have Questions?
Catalog Number
4393711
Unit Size
960 samples
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
384 samples
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
4393711960 samples
-
4393716384 samples
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
3500 Dx Series POP-6™ Polymer is a separation matrix for the 3500 Dx⁄3500xL Dx genetic analyzers. It is conveniently offered in easy-to-use pouch packages in either 960- or 384-sample size formats.

Easy to Use
POP polymers are pre-formulated in a single-use pouch, which not only saves time but also ensures reliability and consistency.

Radio Frequency Identification (RFID) Label
RFID labels encode easy-to-track information such as part⁄lot number, samples remaining, and expiration dates (displayed on software dashboard).

Robust Formulation
POP polymers are formulated for a wide range of applications.

Reproducible Results
POP™ polymers dynamically coat the capillary wall to control electro-osmotic flow. Their defined quality and uniform consistency eliminate guesswork and help to ensure reproducibility. POP polymers are specifically formulated to separate DNA fragments of a known size range at a desired resolution and run time.

Performance Optimized Polymer
This product is intended for in-vitro diagnostic use on the Applied Biosystems 3500 Dx Series Genetic Analyzers and is not validated for use with other systems or applications. Refer to the Applied Biosystems™ 3500 Dx⁄3500xL Dx Genetic Analyzer User Guide for further details.

Specifications

Capillary Length
50 cm, 36 cm
For Use With (Equipment)
3500xL Dx Genetic Analyzer, 3500 Dx Genetic Analyzer
For Use With (Application)
Fragment Analysis, Standard Sequencing
On Instrument Life Usage
Contains enough polymer for 960 samples; polymer is stable on instrument for 7 days
Product Line
POP-6™
Product Type
Polymer
Quantity
960 Samples
Shipping Condition
Wet Ice
Test Time
2 hrs for 50 cm capillary (long read), 1 hr for 36 cm capillary (std. seq.)

Contents & Storage

Store the polymer pouch at 2-8°C until ready to use.
Note that expired pouches cannot be used on the instrument.

On-instrument life⁄usage

3500 Dx (8 capillary) Genetic Analyzer: lesser of 7 days or 384 samples or 60 injections

3500xL Dx (24 capillary) Genetic Analyzer: lesser of 7 days or 384 samples or 20 injections

The pouch includes additional polymer volume to accommodate both installation and a reasonable number of post-installation wizard operations such as bubble removal without affecting the number of samples⁄injections available.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text