Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Recommended Storage | The pouch contains 384 samples of the polymer. Store the pouch at 2-8°C until ready to use. Refer to the expiration date on the label. Note that expired pouches cannot be used on the instrument. |
Quantity | 384 Samples |
For Use With (Application) | Human Identification |
Product Line | POP-4™ |
Unit Size | 384 samples |
Recommended Storage | The pouch contains 96 samples of the polymer. Store pouch at 2–8°C until ready to use. Refer to the expiration date on the label. Note that expired pouches should not be used on the instrument. |
Quantity | 96 Samples |
For Use With (Application) | Human Identification |
Product Line | POP-4™ |
Unit Size | 96 samples |
Recommended Storage | The pouch contains 960 samples of the polymer. Store the pouch at 2-8°C until ready to use. Refer to the expiration date on the label. Note that expired pouches cannot be used on the instrument. |
Quantity | 960 Samples |
For Use With (Application) | Human Identification |
Product Line | POP-4™ |
Unit Size | 960 samples |
Catalog Number | Specifications | Unit Size | Quantity | Price (USD) | Availability | Quantity | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
4393715 | 384 samples | 384 Samples | 267.65 Online price 292.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
A26070 | 96 samples | 96 Samples | 92.65 Online price 97.50 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
4393710 | 960 samples | 960 Samples | 648.65 Online price 732.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
POP-4, POP-6, and POP-7 are conveniently offered in easy to use pouch packages with multiple sample size formats. The POP-4 separation matrix is optimized for HID/forensic applications.
• Easy to use: Pre-formulated and in a single use pouch, which not only saves time but also ensures reliability and consistency
• Include radio frequency identification (RFID) label: easy to track important information such as part/lot number, samples remaining, and expiry dates (displayed on software dashboard)
• Robust formulation: formulated for sequencing and fragment analysis applications
• Replenishable: alow capillaries to be used multiple times
Reproducible Results
POP polymers dynamically coat the capillary wall to control electro-osmotic flow. Their definite quality and uniform consistency eliminate guesswork and help to ensure reproducibility. POP Polymers are specifically formulated to separate DNA fragments of a known size range at a desired resolution and run time.
Many Choices
POP-7 polymers are very versatile and designed for a wide range of short read to long read sequencing applications as well as fragment analysis. POP-4 polymers are designed specifically for HID/Forensic applications, while POP-6 polymers are the choice for certain sequencing applications (and fragment analysis).
For partially used pouches that need to be stored, a cap (4412619) that plugs into the pouch fitment opening is recommended.