Anode Buffer Container for 3500 Dx/3500xL Dx Genetic Analyzers
Applied Biosystems™
Anode Buffer Container for 3500 Dx/3500xL Dx Genetic Analyzers
This is an anode buffer container designed for use with the 3500 Dx⁄3500xL Dx Genetic Analyzer.The 3500 Dx series conditioningRead more
Have Questions?
Catalog number 4393925
Price (USD)/ 4 each
191.00
-
Add to cart
Price (USD)/ 4 each
191.00
Add to cart
Anode Buffer Container for 3500 Dx/3500xL Dx Genetic Analyzers
Catalog number4393925
Price (USD)/ 4 each
191.00
-
Add to cart
This is an anode buffer container designed for use with the 3500 Dx⁄3500xL Dx Genetic Analyzer.
The 3500 Dx series conditioning reagent, anode buffer, cathode buffer and septa cathode buffer are required for instrument operation.
The anode buffer container (ABC) contains 1x running buffer to support all electrophoresis applications on the 3500 Dx series Genetic Analyzers. It is made in a ready-to-use, disposable container with a radio frequency identification (RFID) tag incorporated into the label. The top of the ABC is heat sealed with a plastic film, which should be removed prior to direct installation on to the instrument. Each package includes four individual containers.
This product is intended for in-vitro diagnostic use on the Applied Biosystems 3500 Dx Series Genetic Analyzers and is not validated for use with other systems or applications. The 3500 Dx Series Genetic Analyzer is to be used by trained operators in the analysis of human DNA or RNA, for the detection of genetic changes that may lead to disease presence or susceptibility. Off-label uses such as fragment analysis are for Research Use Only. Refer to the Applied Biosystems 3500 Dx⁄3500xL Dx Genetic Analyzer User Guide for further details.
For in-vitro diagnostic use
Specifications
For Use With (Equipment)3500xL Dx Genetic Analyzer, 3500 Dx Genetic Analyzer
For Use With (Application)Sequencing
Product TypeAnode Buffer Container
Quantity4 each
Shipping ConditionWet Ice
Unit Size4 each
Contents & Storage
Each package contains 4 buffer containers. Store at 2 - 8°C.