2-Chloro-1,3-dimethylimidazolinium chloride, 90%, Thermo Scientific Chemicals
2-Chloro-1,3-dimethylimidazolinium chloride, 90%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Chloro-1,3-dimethylimidazolinium chloride, 90%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Packaging:
Glass bottle
Catalog number 439521000
Price (USD)/ Each
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS37091-73-9
IUPAC Name2-chloro-1,3-dimethyl-4,5-dihydro-1H-imidazol-3-ium chloride
Molecular FormulaC5H10Cl2N2
InChI KeyAEBBXVHGVADBHA-UHFFFAOYSA-M
SMILES[Cl-].CN1CC[N+](C)=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Elemental Analysis31.26 to 39.79 % (C)
Appearance (Form)Crystalline powder or solid
HPLC>=88.0 %
Elemental Analysis14.58 to 18.56 % (N)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Coupling reagent
RUO – Research Use Only