Thermo Scientific Chemicals

1,1'-Oxalyldiimidazole, Technical, Thermo Scientific Chemicals

Catalog number: 439620050
5 g, Each
Thermo Scientific Chemicals

1,1'-Oxalyldiimidazole, Technical, Thermo Scientific Chemicals

Catalog number: 439620050
5 g, Each
Quantity
Catalog number: 439620050
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
18637-83-7
IUPAC Name
bis(1H-imidazol-1-yl)ethane-1,2-dione
Molecular Formula
C8H6N4O2
InChI Key
ONRNRVLJHFFBJG-UHFFFAOYSA-N
SMILES
O=C(N1C=CN=C1)C(=O)N1C=CN=C1
Appearance (Color)
Yellow to brown
Appearance (Form)
Powder
Infrared spectrum
Conforms
HPLC
>=80.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Coupling reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text