Thermo Scientific Chemicals

6-Quinolineboronic acid pinacol ester, 97%, Thermo Scientific Chemicals

Catalog number: 439640010
1 g, Each
Thermo Scientific Chemicals

6-Quinolineboronic acid pinacol ester, 97%, Thermo Scientific Chemicals

Catalog number: 439640010
1 g, Each
Quantity
Catalog number: 439640010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
406463-06-7
IUPAC Name
6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)quinoline
Molecular Formula
C15H18BNO2
InChI Key
VMFALDWCEQUFSX-UHFFFAOYSA-N
SMILES
CC1(C)OB(OC1(C)C)C1=CC2=CC=CN=C2C=C1
Melting point
62°C to 70°C
Appearance (Color)
White to light orange
Appearance (Form)
Powder or crystalline solid
Infrared spectrum
Conforms
GC
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text