TaqMan™ Environmental Master Mix 2.0
TaqMan™ Environmental Master Mix 2.0
Applied Biosystems™

TaqMan™ Environmental Master Mix 2.0

Applied Biosystems™ TaqMan™ Environmental Master Mix 2.0 offers accurate, real-time PCR-based pathogen detection in the presence of high levels ofRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNo. of Reactions
4396838200 Reactions
4398021400 Reactions
4398044800 Reactions
Catalog number 4396838
Price (EUR)
738,00
Each
-
Add to cart
No. of Reactions:
200 Reactions
Recurring order eligible. Learn more »
Price (EUR)
738,00
Each
Add to cart
Ask our AI about this Product
Applied Biosystems™ TaqMan™ Environmental Master Mix 2.0 offers accurate, real-time PCR-based pathogen detection in the presence of high levels of inhibitors. Use this Master Mix to analyze environmental, food and other challenging samples.

- Tailored for quantitative, real-time PCR experiments
- Unrivalled sensitivity for challenging applications, low copy number target detection and multiplex PCR
- Includes pathogen detection and viral⁄bacterial load quantification
- Sensitive detection for reliable quantification of abundant and limited genetic targets
- Stable mix for high-throughput handling

Optimized Formulation for Unrivalled Performance

TaqMan™ Environmental Master Mix 2.0 is a convenient 2X mix for target quantification that includes:

- AmpliTaq Gold™ DNA Polymerase, UP (Ultra Pure), a highly purified DNA Polymerase for improved detection of bacterial targets. This hot-start enzyme is inactive at room temperature so reactions can be set up on the benchtop. Enzyme is activated during thermal cycling.
- Passive internal reference based on proprietary ROX™ dye for precise data analysis.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Concentration2X
For Use With (Equipment)7000 System, 7300 System, 7500 Fast System, 7500 System, 7900HT Fast System, 7900HT System, StepOne™, Standard Mode
FormatBottle
Hot StartBuilt-In Hot Start
No. of Reactions200 Reactions
Passive Reference DyeROX (Pre-mixed)
PolymeraseAmpliTaq Gold DNA Polymerase
Product LineTaqMan™
Product TypeqPCR Master Mix
Purity or Quality GradeUP (Ultra Pure)
Quantity200 reactions
Sample TypeEnvironmental Sample, Food
Shipping ConditionDry Ice
Sufficient For200 Reactions
Volume5 mL
Detection MethodPrimer-probe
For Use With (Application)Gene Expression, Genotyping, Pathogen Detection, Virus Detection
PCR MethodqPCR
Reaction SpeedStandard
Unit SizeEach
Contents & Storage
This TaqMan Environmental Master Mix 2.0 is a convenient 2X mix in a 5 mL bottle that includes: AmpliTaq Gold DNA Polymerase, UP (Ultra Pure), and a passive internal reference based on proprietary ROX dye for precise data analysis.
Upon receipt, store at -15 to -25°C before first use, protected from light. After first use, store at 2 to 8°C, protected from light.
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
2504182Certificate of AnalysisJun 14, 20254396838
2412180Certificate of AnalysisMay 27, 20254396838
2502181Certificate of AnalysisMar 16, 20254396838
2410179Certificate of AnalysisOct 30, 20244396838
2407178Certificate of AnalysisAug 05, 20244396838
5 results displayed, search above for a specific certificate

Safety Data Sheets

Frequently asked questions (FAQs)

No, the 2X Environmental Master Mix (EMM) is not the same as the TaqMan Universal PCR Master Mix (with or without AmpErase UNG). The 2X Environmental Master Mix is specifically formulated to detect specific bacterial pathogens with greater specificity and sensitivity. It includes AmpliTaq Gold DNA Polymerase, LD, which is a highly purified version of AmpliTaq Gold DNA Polymerase. Every effort has been made in the development of this new polymerase to eliminate bacterial DNA from the enzyme preparation.

Yes, it is.

Find additional tips, troubleshooting help, and resources within our Gene Expression Analysis & Genotyping Support Center.

Yes. You can purchase the TaqMan Environmental Master Mix 2.0 (Cat. No. 4396838).

Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center

It is sufficient for 100 PCR reactions.

For fecal samples, we recommend a master mix that is very tolerant of inhibitors, such as our TaqMan Environmental Master Mix 2.0. Please refer to the following paper: https://assets.thermofisher.com/TFS-Assets/CMD/posters/cms_089094.pdf

Share catalog number, name or link

1x1 image pixel for data collection