1-Isobutyl-1H-pyrazole-4-boronic acid pinacol ester, 97%
1-Isobutyl-1H-pyrazole-4-boronic acid pinacol ester, 97%
1-Isobutyl-1H-pyrazole-4-boronic acid pinacol ester, 97%
1-Isobutyl-1H-pyrazole-4-boronic acid pinacol ester, 97%
Thermo Scientific Chemicals

1-Isobutyl-1H-pyrazole-4-boronic acid pinacol ester, 97%

CAS: 827614-66-4 | C13H23BN2O2 | 250.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 439730050
Price (EUR)
176,80
Online Exclusive
208,00
Save 31,20 (15%)
Each
-
Add to cart
Quantity:
5 g
Recurring order eligible. Learn more »
Request bulk or custom format
Price (EUR)
176,80
Online Exclusive
208,00
Save 31,20 (15%)
Each
Add to cart
Chemical Identifiers
CAS827614-66-4
IUPAC Name1-(2-methylpropyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole
Molecular FormulaC13H23BN2O2
InChI KeyYMEBZRNYQBODKB-UHFFFAOYSA-N
SMILESCC(C)CN1C=C(C=N1)B1OC(C)(C)C(C)(C)O1
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=96.0 %
Appearance (Color)Colorless to yellow
Appearance (Form)Liquid
Infrared spectrumConforms
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Safety Data Sheets

Safety and Handling

Classification of the substance or mixture
CLP classification - Regulation(EC) No 1272/2008
Flammable liquids
Category 3
Skin Corrosion/Irritation
Category 2
Serious Eye Damage/Eye Irritation
Category 2
Specific target organ toxicity - (single exposure)
Category 3
Label Elements
Signal Word

Warning

Hazard Statements

H315 - Causes skin irritation

H319 - Causes serious eye irritation

H335 - May cause respiratory irritation

Physical Hazards

H226 - Flammable liquid and vapor

Precautionary Statements

P210 - Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking

P264 - Wash face, hands and any exposed skin thoroughly after handling

P280 - Wear protective gloves/protective clothing/eye protection/face protection

P303 + P361 + P353 - IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water or shower

P304 + P340 - IF INHALED: Remove person to fresh air and keep comfortable for breathing

P312 - Call a POISON CENTER or doctor if you feel unwell

P332 + P313 - If skin irritation occurs: Get medical advice/attention

P337 + P313 - If eye irritation persists: Get medical advice/attention

Share catalog number, name or link

1x1 image pixel for data collection