Specifications
Detection MethodPrimer-probe
FormatBottle
PCR MethodqPCR
PolymeraseAmpliTaq Gold DNA Polymerase
Reaction SpeedStandard
For Use With (Equipment)7000 System, 7300 System, 7500 Fast System, 7500 System, 7900HT Fast System, 7900HT System, StepOne™, Standard Mode
Passive Reference DyeROX (Pre-mixed)
Product LineTaqMan™
Product TypeqPCR Master Mix
Purity or Quality GradeUP (Ultra Pure)
Sufficient For400 Reactions
For Use With (Application)Gene Expression, Genotyping, Pathogen Detection, Virus Detection
Concentration2X
Hot StartBuilt-In Hot Start
No. of Reactions400 Reactions
Sample TypeEnvironmental Sample, Food
Volume10 mL
Unit Size400 reactions