AmpliTaq™ 360 DNA Polymerase
Inquire about OEM or Commercial Supply version of this product here.
Switch to Thermo Scientific DreamTaq DNA Polymerases for better results and added convenience.
AmpliTaq™ 360 DNA Polymerase
Applied Biosystems™

AmpliTaq™ 360 DNA Polymerase

Green features
The Applied Biosystems AmpliTaq360 DNA Polymerase, when used with the new enhanced AmpliTaq 360 Buffer and the optional 360 GC Enhancer, amplifies a vast range of DNA sequence contexts.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
4398818250 Units
43988281000 Units
439889725 x 1000 Units
43988955000 Units
Catalog number 4398818
Price (USD)/ Each
193.00
-
Add to cart
Quantity:
250 Units
Request bulk or custom format
Price (USD)/ Each
193.00
Add to cart
AmpliTaq™ 360 DNA Polymerase
Catalog number4398818
Price (USD)/ Each
193.00
-
Add to cart
The Applied Biosystems AmpliTaq360 DNA Polymerase, when used with the new enhanced AmpliTaq 360 Buffer and the optional 360 GC Enhancer, amplifies a vast range of DNA sequence contexts. Compared to the original AmpliTaq DNA Polymerase, AmpliTaq 360 DNA Polymerase is purified by an additional proprietary separation process that reduces contaminating bacterial DNA sequences from the enzyme preparation. This ultra-pure enzyme reduces false positive results, amplifies low-level target sequences, and, when combined with the proprietary AmpliTaq 360 Buffer Kit, promotes the amplification of a variety of templates, including those from bacterial and human genomes.

Features

  • Optimized for the broadest range of targets, from everyday to challenging
  • Unmatched sensitivity and yield
  • Market-leading GC enhancer for robust amplification of GC-rich sequences
  • Achieves the highest quality sequencing data

Notes

  • DreamTaq DNA Polymerase is recommended for superior PCR performance.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
FormatStand-alone enzyme
GC-Rich PCR PerformanceMedium
PolymeraseAmpliTaq 360 DNA Polymerase
Reaction SpeedStandard
Exonuclease Activity5' - 3'
Green FeaturesSustainable packaging
Product TypeDNA Polymerase
Quantity250 Units
Shipping ConditionRoom Temp or Wet Ice
For Use With (Application)Standard PCR
Concentration5 U/μL
Fidelity (vs. Taq)1X
Hot StartNo
No. of Reactions200 Reactions
Overhang3'-A
Reaction FormatSeparate Components
Size (Final Product)5 kb or less
Starting MaterialDNA
Unit SizeEach
Contents & Storage
• AmpliTaq 360 DNA Polymerase (5 U/μL), 50 μL
• 10X AmpliTaq 360 PCR Buffer, 1.5 mL
• 25 mM MgCl2, 1.5 mL
• 360 GC Enhancer, 1.5 mL

Store at -15°C to -25°C.
Product is guaranteed up to 12 months as indicated by the expiration date.