Informational:Inquire about OEM or Commercial Supply version of this product here.
Informational:Upgrade to Platinum II Taq Hot-Start DNA Polymerase for universal annealing and a 4X faster cycling protocol.
Applied Biosystems™

AmpliTaq Gold™ 360 Master Mix

Catalog number: 4398886
Have Questions?
Applied Biosystems™

AmpliTaq Gold™ 360 Master Mix

Catalog number: 4398886
Have Questions?
Catalog Number
4398886
Unit Size
50 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
5 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
1 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
10 x 5 mL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
439888650 mL
-
43988815 mL
-
43988761 mL
-
439890110 x 5 mL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
AmpliTaq Gold™ 360 PCR Master Mix contains everything required for successful PCR amplification in one convenient package—all components are premixed and premeasured. AmpliTaq Gold™ 360 Master Mix was designed for 360° coverage of a full range of targets. The master mix is supplied at 2X the recommended usage concentration for easy dilution when adding template and primers. Designed for convenience, the AmpliTaq Gold™ 360 PCR Master Mix scales to various reaction volumes for greater application and format flexibility. The main ingredient of the master mix is AmpliTaq Gold™ 360 DNA Polymerase but it also contains the world-class 360 GC Enhancer which can be added optionally for high GC–content templates. With the wide template coverage, optimization of PCR reaction conditions, is virtually eliminated with the use of AmpliTaq Gold™ 360 Master Mix. Key features:

• Optimized for the broadest range of targets—from everyday to challenging
• Unmatched sensitivity, specificity, and yield
• Robust amplification of GC-rich sequences with market-leading 360 GC Enhancer
• Achieves the highest-quality sequencing data
• Easy-to-use, premixed master mix

Optimized for easy and challenging targets
Challenging targets include AT-rich, GC-rich, primer-dimer–forming amplicons, homopolymer repeats, and amplicons that pose sequencing challenges. Amplicons that previously required specialized enzymes and reaction conditions can now be reproducibly amplified with a single reagent under standardized conditions (see figure). Competitive benchmarking across more than 40 amplicons distinguishes AmpliTaq Gold™ 360 as the best-performing enzyme, ensuring the highest probability of success for the amplification of both everyday and challenging targets (see table).

Optimized for automated, hot-start PCR
AmpliTaq Gold™ 360 DNA Polymerase is the key ingredient in an automated, convenient, and efficient hot-start PCR. When AmpliTaq Gold™ 360 PCR Master Mix is added to the reaction mixture at room temperature, the inactive enzyme is not capable of primer extension. Any low-stringency mispriming events that may have occurred will not be enzymatically extended and subsequently amplified. An initial thermal incubation step is required for activation and ensures that active enzyme is generated only at temperatures where the DNA is fully denatured. The amount of AmpliTaq Gold™ 360 DNA polymerase increases in the reaction slowly with each cycle number, and specific product yield increases without buildup of nonspecific products, including primer dimers. Excellent specificity across a broad range of targets (see figure). The extreme specificity allows easier multiplexing and allelic discrimination.

Amplify low-copy amplicons and long targets
AmpliTaq Gold™ 360 DNA Polymerase efficiently amplifies targets present at low copy number (see figure), even in the presence of high concentrations of complex DNA, making it especially suited for low-copy pathogen detection, and amplification of targets from degraded DNA samples. The extreme purity of the enzyme contributes to its unmatched sensitivity. AmpliTaq Gold™ 360 DNA Polymerase efficiently and reproducibly amplifies long (up to 5 kb) sequences. The figure shown demonstrates robust PCR amplification of long human and plasmid DNA.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Detection Method
Primer-probe
Format
Tube
GC-Rich PCR Performance
High
Polymerase
AmpliTaq Gold 360 DNA Polymerase
Reaction Speed
Standard
Exonuclease Activity
5' - 3'
Green Features
Sustainable packaging
Product Line
AmpliTaq Gold™
Product Type
PCR Master Mix
Quantity
50 mL
Shipping Condition
Approved for shipment at Room Temperature or on Wet Ice
For Use With (Application)
Cloning
Concentration
2X
Fidelity (vs. Taq)
1 X
Hot Start
Built-In Hot Start
No. of Reactions
2000
Overhang
3'-A
Reaction Format
SuperMix or Master Mix
Size (Final Product)
5 kb or less
Starting Material
DNA
Volume
50 mL

Contents & Storage

• 1 x 50 mL 2X AmpliTaq Gold 360 Master Mix
• 1 x 10 mL 360 GC Enhancer

Store at -20°C until first use, then store at 4°C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text