2-Pyridineboronic acid N-phenyl-diethanolamine ester, 50-70%, see CofA for exact com, Thermo Scientific Chemicals
2-Pyridineboronic acid N-phenyl-diethanolamine ester, 50-70%, see CofA for exact com, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Pyridineboronic acid N-phenyl-diethanolamine ester, 50-70%, see CofA for exact com, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
10 g
Packaging:
Glass bottle
Catalog number 439890050
Price (USD)/ Each
129.00
-
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
129.00
Add to cart
2-Pyridineboronic acid N-phenyl-diethanolamine ester, 50-70%, see CofA for exact com, Thermo Scientific Chemicals
Catalog number439890050
Price (USD)/ Each
129.00
-
Add to cart
Chemical Identifiers
IUPAC Name4-phenyl-8-(pyridin-2-yl)-hexahydro-4λ⁵-[1,3,2]oxazaborolo[2,3-b][1,3,2]oxazaborol-4-ylium-8-uide
Molecular FormulaC15H17BN2O2
InChI KeyFZYLTWAWSRCZPG-UHFFFAOYSA-N
SMILESC1C[N+]2(CCO[B-]2(O1)C1=NC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol)268.12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Powder
Infrared spectrumConforms
NMR (1H-NMR)Consistent with structure
Additional infoContains varying amounts of IPA and
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only