Thermo Scientific Chemicals

2-Pyridineboronic acid N-phenyl-diethanolamine ester, 50-70%, see CofA for exact com, Thermo Scientific Chemicals

Catalog number: 439890100
10 g, Glass bottle, Each
Thermo Scientific Chemicals

2-Pyridineboronic acid N-phenyl-diethanolamine ester, 50-70%, see CofA for exact com, Thermo Scientific Chemicals

Catalog number: 439890100
10 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 439890100
Price (USD)
Have Questions?

Chemical Identifiers

CAS
662138-96-7
IUPAC Name
4-phenyl-8-(pyridin-2-yl)-hexahydro-4λ⁵-[1,3,2]oxazaborolo[2,3-b][1,3,2]oxazaborol-4-ylium-8-uide
Molecular Formula
C15H17BN2O2
InChI Key
FZYLTWAWSRCZPG-UHFFFAOYSA-N
SMILES
C1C[N+]2(CCO[B-]2(O1)C1=NC=CC=C1)C1=CC=CC=C1
Appearance (Color)
White
Appearance (Form)
Powder
Infrared spectrum
Conforms
NMR (1H-NMR)
Consistent with structure
Additional info
Contains varying amounts of IPA and

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text