TaqMan™ Array Human MicroRNA A Cards v2.0
TaqMan™ Array Human MicroRNA A Cards v2.0
Applied Biosystems™

TaqMan™ Array Human MicroRNA A Cards v2.0

The TaqMan® Array Human MicroRNA A Card v2.0 contains 384 TaqMan® MicroRNA Assays enabling accurate quantitation of 377 human microRNAsRead more
Have Questions?
Catalog number 4398965
Price (USD)/ Each
2,750.00
-
Add to cart
Price (USD)/ Each
2,750.00
Add to cart
TaqMan™ Array Human MicroRNA A Cards v2.0
Catalog number4398965
Price (USD)/ Each
2,750.00
-
Add to cart
The TaqMan® Array Human MicroRNA A Card v2.0 contains 384 TaqMan® MicroRNA Assays enabling accurate quantitation of 377 human microRNAs in as little as 5 hours. Three TaqMan MicroRNA Assay endogenous controls are included to aid in data normalization and one TaqMan® MicroRNA Assay not related to human is included as a negative control. Details on the included assays is available in the Target List file. Purchase of the Megaplex™ RT Primers, Human Pool A v2.1 is required to run this array. An additional preamplification step is enabled by Megaplex™ PreAmp Primers, Human Pool A v2.1 for situations where sensitivity is of the utmost importance and⁄or sample is limiting.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
TypeArray Human MicroRNA A Card
Format384-well Microfluidic Card
SpeciesHuman
FormDried Down
For Use With (Equipment)AB 7900 HT 384-Well System
Internal Probe ModificationMGB (Minor Groove Binder) (3'), Non-Fluorescent Quencher (3'), FAM (5')
Label or DyeFAM
Product LineMegaplex™, TaqMan™
Quantity4 pack
Reaction SpeedStandard
Research CategoryGene Regulation, Epigenetics
Unit SizeEach
Contents & Storage
4°C