N-Phenyl-2-naphthylamine, 97%, Thermo Scientific Chemicals
N-Phenyl-2-naphthylamine, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-Phenyl-2-naphthylamine, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 439910250
Price (USD)/ Each
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS135-88-6
IUPAC NameN-phenylnaphthalen-2-amine
Molecular FormulaC16H13N
InChI KeyKEQFTVQCIQJIQW-UHFFFAOYSA-N
SMILESN(C1=CC=CC=C1)C1=CC=C2C=CC=CC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Off-white to grey to brown
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Melting point105°C to 108°C
Titration with HClO4>=96.0 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only