Thermo Scientific Chemicals

N-Phenyl-2-naphthylamine, 97%, Thermo Scientific Chemicals

Catalog number: 439911000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

N-Phenyl-2-naphthylamine, 97%, Thermo Scientific Chemicals

Catalog number: 439911000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 439911000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
135-88-6
IUPAC Name
N-phenylnaphthalen-2-amine
Molecular Formula
C16H13N
InChI Key
KEQFTVQCIQJIQW-UHFFFAOYSA-N
SMILES
N(C1=CC=CC=C1)C1=CC=C2C=CC=CC2=C1
Appearance (Color)
Off-white to grey to brown
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms
Melting point
105°C to 108°C
Titration with HClO4
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text