Applied Biosystems™

Megaplex™ RT Primers, Human Pool A v2.1

Catalog number: 4399966
Have Questions?
Applied Biosystems™

Megaplex™ RT Primers, Human Pool A v2.1

Catalog number: 4399966
Have Questions?
Catalog Number
4399966
Unit Size
50 reactions
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
439996650 reactions
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Designed to streamline experimental workflows, Megaplex™ RT Primers provide a single reaction solution when preparing cDNA for real-time PCR analysis on a TaqMan® MicroRNA Array. Human Pool A v2.1 contains RT primers for 377 unique microRNAs and 4 controls. The microRNA targets represented in this pool tend be better characterized, more broadly expressed and⁄or expressed at a higher level compared to Human Pool B. Human Pool A v2.1 is intended for use with the TaqMan® Array, Human MicroRNA A Card v2.0, however, it can also be used to prepare cDNA for real-time analysis using the individual TaqMan® MicroRNA Assays.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

GC-Rich PCR Performance
High
PCR Method
qPCR
Green Features
Sustainable packaging
Primer
RT Primers
Product Line
Megaplex™, TaqMan™
Product Type
RT Primer
Shipping Condition
Room Temperature
Species
Human
Sufficient For
50 Reactions
For Use With (Application)
miRNA analysis
No. of Reactions
50 Reactions

Contents & Storage

Store at -20°C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text