Applied Biosystems™

Megaplex™ RT Primers, Rodent Pool A

Catalog number: 4399970
Have Questions?
Applied Biosystems™

Megaplex™ RT Primers, Rodent Pool A

Catalog number: 4399970
Have Questions?
Catalog Number
4399970
Unit Size
50 reactions
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
439997050 reactions
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Designed to streamline experimental workflows, Applied Biosystems® Megaplex™ RT Primers provide a single reaction solution when preparing cDNA for real-time PCR analysis on a TaqMan® MicroRNA Array. Rodent Pool A contains RT primers for 335 and 238 unique microRNAs for mouse and rat respectively plus 4 species specific controls. The microRNA targets represented in this pool tend be better characterized, more broadly expressed and⁄or expressed at a higher level compared to Human Pool A. The pool can also be used to prepare cDNA for real-time analysis using the individual TaqMan® MicroRNA Assays.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

GC-Rich PCR Performance
High
PCR Method
qPCR
Green Features
Sustainable packaging
Primer
RT Primers
Product Line
Megaplex™, TaqMan™
Product Type
RT Primer
Shipping Condition
Room Temperature
Species
Rat, Mouse
Sufficient For
50 Reactions
Concentration
10X
Fidelity (vs. Taq)
10 X
No. of Reactions
50 Reactions
Volume
50 μL

Contents & Storage

Contains 50μL of 10X primers and 1 mL 25 mM MgCl2.

Store at -15° to -25°C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text