1-Piperidinepropionic acid, 96%, Thermo Scientific Chemicals
1-Piperidinepropionic acid, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-Piperidinepropionic acid, 96%, Thermo Scientific Chemicals

Have Questions?
Quantity:
25 g
Catalog number 440050250
Price (USD)/ Each
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS26371-07-3
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
HPLC>=95.0 %
Appearance (Color)White to light brown
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only