Thermo Scientific Chemicals

Poly(methylhydrosiloxane), trimethylsilyl terminated, Thermo Scientific Chemicals

Catalog number: 440100250
25 mL, Each
Thermo Scientific Chemicals

Poly(methylhydrosiloxane), trimethylsilyl terminated, Thermo Scientific Chemicals

Catalog number: 440100250
25 mL, Each
Quantity
Catalog number: 440100250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
63148-57-2
Molecular Formula
(CH4OSi)n
InChI Key
QNWOFLWXQGHSRH-UHFFFAOYSA-N
SMILES
C[SiH](O[Si](C)(C)C)O[Si](C)(C)C
MDL Number
MFCD00084478
Viscosity
15 to 25 cSt
Infrared spectrum
Conforms
Appearance (Color)
Colorless
Appearance (Form)
Liquid
Refractive index
1.3960 to 1.4000 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text