Poly(methylhydrosiloxane), trimethylsilyl terminated, Thermo Scientific Chemicals
Poly(methylhydrosiloxane), trimethylsilyl terminated, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Poly(methylhydrosiloxane), trimethylsilyl terminated, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
25 mL
Catalog number 440101000
Price (USD)/ Each
-
Quantity:
100 mL
Request bulk or custom format
Chemical Identifiers
Molecular Formula(CH4OSi)n
InChI KeyQNWOFLWXQGHSRH-UHFFFAOYSA-N
SMILESC[SiH](O[Si](C)(C)C)O[Si](C)(C)C
MDL NumberMFCD00084478
CAS63148-57-2
SpecificationsSpecification SheetSpecification Sheet
Viscosity15 to 25 cSt
Infrared spectrumConforms
Appearance (Color)Colorless
Appearance (Form)Liquid
Refractive index1.3960 to 1.4000 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only