Applied Biosystems™

TaqMan™ GTXpress™ Master Mix

Catalog number: 4401890
Have Questions?
Applied Biosystems™

TaqMan™ GTXpress™ Master Mix

Catalog number: 4401890
Have Questions?
Catalog Number
4401890
Unit Size
50 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
1 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
10 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
2 x 50 mL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
440189050 mL
-
44033111 mL
-
440189210 mL
-
44018572 x 50 mL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
**********************
Alternative Product: Try TaqPath ProAmp Master Mix, our newest and highest performing genotyping master mix for accurate results, even from samples containing PCR inhibitors. With TaqPath ProAmp Master Mix, we’ve taken the best of TaqMan GTXpress Master Mix and added additional capabilities for your genotyping experiments.

**********************
TaqMan™ GTXpress™ Master Mix delivers accurate genotyping results with robust performance in less than 50 min. Applied Biosystems™ TaqMan™ GTXpress™ Master Mix (2X) combines AmpliTaq™ Fast DNA Polymerase, UP, dNTPs, Tracking Dye, and ROX™ dye in a convenient single vial format. The 1 mL pack includes reagents sufficient to perform 400 PCR reactions (5 μL/reaction).
• Compatible with both Fast and Standard mode thermal cycling conditions
• Pre-mixed components stored at 2°–8°C significantly reduce assay setup time
• Validated with all types of Applied Biosystems™ TaqMan™ Genotyping Assays
• Extended PCR cycling can be used for poorly amplified or sample-limited reactions The TaqMan™ GTXpress™ Master Mix is also sold as part of TaqMan™ Sample-to-SNP™ Kit, which provides real-time PCR-ready DNA in 5 min from a wide variety of samples, ranging from blood to buccal swabs to plant tissues. For more information on the TaqMan™ Sample-to-SNP™ Kit, refer to the Applied Biosystems™ TaqMan™ Sample-to-SNP™ Kit Protocol (PN. 4402136).
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Detection Method
Primer-probe
Format
Single Vial
GC-Rich PCR Performance
High
PCR Method
qPCR
Polymerase
AmpliTaq DNA Polymerase
Reaction Speed
Fast or Standard
Technique
SNP Genotyping (Real-Time PCR-Based)
For Use With (Equipment)
QuantStudio™ 12k Flex, QuantStudio™ 3, QuantStudio™ 5, QuantStudio™ 6 Flex, QuantStudio™ 7, StepOne™ Standard Mode, StepOne™ Fast Mode
Genotyping Target
SNPs (Unknown or Numerous), SNPs (Known)
Passive Reference Dye
ROX (Pre-mixed)
Product Line
GTXpress™, TaqMan™
Product Type
Master Mix
Purity or Quality Grade
UP (Ultra Pure)
Quantity
50 mL
Shipping Condition
Wet Ice
For Use With (Application)
Genotyping
Concentration
2X
Fidelity (vs. Taq)
2 X
Hot Start
Built-In Hot Start
No. of Reactions
40,000 Reactions
Sample Type
DNA (Genomic), Sample-to-SNP™ Kit Lysates
Volume
100 mL

Contents & Storage

Contains 1 x 50 mL TaqMan™ GTXpress™ Master Mix (2X master mix with AmpliTaq™ Fast DNA Polymerase, UP, dNTPs, Tracking Dye and ROX™ Dye). Sufficient for 20,000 reactions. Store at 2-8°C.

Guaranteed minimum shelf life is 60 days (exact expiry date printed on product and CofA).

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text