Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Detection Method | Primer-probe |
Format | Kit |
PCR Method | qPCR |
Host Cell | CHO |
Label or Dye | FAM |
Product Line | resDNASEQ™ |
Shipping Condition | Dry Ice |
No. of Reactions | 100 Reactions |
Includes | Assay Kit only |
Unit Size | 100 reactions |
Detection Method | Primer-probe |
Format | Kit |
PCR Method | qPCR |
Host Cell | CHO |
Label or Dye | FAM |
Product Line | resDNASEQ™ |
No. of Reactions | 100 Reactions |
Includes | Assay Kit and Sample Preparation |
Unit Size | 100 reactions |
Catalog Number | Specifications | Unit Size | Includes | Price (USD) | Availability | Quantity | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
4402085 | 100 reactions | Assay Kit only | 3,455.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
4413713 | 100 reactions | Assay Kit and Sample Preparation | 4,035.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|