Thermo Scientific Chemicals

Potassium sulfite, 90%, pure, Thermo Scientific Chemicals

Catalog number: 440210025
2.5 kg, Glass Bottle, Each
Thermo Scientific Chemicals

Potassium sulfite, 90%, pure, Thermo Scientific Chemicals

Catalog number: 440210025
2.5 kg, Glass Bottle, Each
Quantity
Packaging
Catalog number: 440210025
Price (USD)
Have Questions?

Chemical Identifiers

CAS
10117-38-1
IUPAC Name
dipotassium sulfite
Molecular Formula
K2O3S
InChI Key
BHZRJJOHZFYXTO-UHFFFAOYSA-L
SMILES
[K+].[K+].[O-]S([O-])=O
Appearance (Form)
Crystalline powder
Titration Iodimetric
>=88 %
Appearance (Color)
White

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text