Applied Biosystems™

DNA Extract All Reagents Kit

Catalog number: 4402599
Have Questions?
Applied Biosystems™

DNA Extract All Reagents Kit

Catalog number: 4402599
Have Questions?
Catalog Number
4402599
Unit Size
200 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
5 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
20 mL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
4402599200 mL
-
44033195 mL
-
440261620 mL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The DNA Extract All Reagents Kit provides PCR-ready DNA from a wide variety of sample types, ranging from blood to buccal swabs to plant tissues in 5 minutes.

Simply mix your sample with the DNA lysis solution provided. Incubate for 3 minutes and add the DNA stabilizing solution, also provided.

After extraction, robust PCR amplification can be completed using TaqMan™ GTXpress™ Master Mix or TaqMan™ OpenArray™ Genotyping Master Mix (pre-amplified sample only), depending on your application. PCR can be performed using either Fast or Standard mode thermal cycling conditions. The DNA Extract All Reagents Kit is included in the TaqMan Sample-to-SNP Kit, so workflow details can be found in the TaqMan™ Sample-to-SNP user guide below.

This 5 mL size should be sufficient for 100 extractions.

Note: Thermal cycling conditions should be followed according to the master mix being used. TaqMan™™ GTXpress™ Master Mix is required for PCR amplification of sample prepared using the DNA Extract All Reagents Kit. If sample has undergone preamplification, OpenArray™ Genotyping Master Mix may also be used.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Product Line
Extract All
Quantity
200 mL
Reagent Type
Cell Lysis Buffer
Test Time
5 min
Form
Liquid
Unit Size
200 mL

Contents & Storage

Store at 2–8°C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text