Informational:Inquire about OEM or Commercial Supply version of this product here.
Invitrogen™

Fast SYBR™ Green Cells-to-CT™ Kit

Catalog number: 4402957
Have Questions?
Invitrogen™

Fast SYBR™ Green Cells-to-CT™ Kit

Catalog number: 4402957
Have Questions?
Catalog Number
4402957
Unit Size
400 reactions
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
100 reactions
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
4402957400 reactions
-
4402956100 reactions
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations

Ambion™ Fast SYBR™ Green Cells-to-CT™ Kit is a fast, easy, and robust method to go directly from cultured cells to real-time PCR results. A breakthrough cell lysis and RNA stabilization technology eliminates the laborious and time-consuming RNA purification process completely. The kit includes sufficient reagents for 400 reactions. The Fast SYBR™ Green Cells-to-CT™ Kit contains AmpliTaq™ Fast DNA Polymerase, UP, which allows instant hot start, thus minimizing non-specific product formation with fast real-time PCR.

• Includes optimized reagents to work efficiently, right out of the box
• Simple, effective Cells-to-CT™ technology enables sample preparation at room temperature in only 7 minutes, including DNase treatment
• Validated accuracy, exceptional reproducibility, and maximal sensitivity from 10 to 100,000 cells per sample; results equivalent to those from purified RNA

Extraordinary Value
The kit includes reagents for sample preparation integrated into a complete, optimized gene expression workflow including reverse transcription reagents and a high-performance PCR master mix with SYBR™ Green.

Extraordinary Ease
The protocol begins with a simple 7-minute sample preparation procedure. First, cells are washed in PBS, and then lysed for 5 minutes at room temperature; DNase treatment can be performed concurrently. The lysis reaction is stopped, and the sample is then ready for reverse transcription or storage at –20°C for up to 5 months. Because samples are processed directly in culture plates (96- or 384-well), sample handling and the potential for sample loss or transfer error are minimized, resulting in higher reproducibility. Following sample preparation, a portion of the cell lysate is added to an RT reaction followed by real-time PCR using the included master mix.

Extraordinary Performance
To demonstrate the performance of the kit, replicate experiments were run using either traditional RNA purification and real-time PCR analysis, or the FastSYBR™ Green Cells-to-CT™ Kit. The kit shows equivalent performance to results obtained using purified RNA for analysis of 10 to 100,000 cells. In addition, the sensitivity, efficiency, and dynamic range were shown to be superior to competitor lysate kits. Samples prepared with the kit have been tested for compatibility on various real-time PCR platforms, including those from Applied Biosystems™, Roche, and Bio-Rad.

Accessory Product:
The SYBR™ Green Cells-to-CT™ Control Kit (Cat. No. 4402959) contains a Xeno™ RNA control and primer set and is recommended to monitor the efficiency of cell lysis and amplification inhibition in Cells-to-CT™ reactions.

WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Detection Method
SYBR
Polymerase
AmpliTaq Fast DNA Polymerase
Reaction Speed
Fast
For Use With (Equipment)
7500 Fast System, 7900HT Fast System, StepOne™, Fast Mode, StepOnePlus™, Fast Mode
Green Features
Fewer resources used and less waste, Less hazardous
Label or Dye
SYBR Green
Passive Reference Dye
ROX (Pre-mixed)
Product Line
Cells-to-CT™, SYBR™
Purification Time
5 min.
Purity or Quality Grade
UP (Ultra Pure)
Reverse Transcriptase
M-MLV
Shipping Condition
Dry Ice
For Use With (Application)
RT-PCR
Concentration
20X
Hot Start
Built-In Hot Start
No. of Reactions
400 reactions
Sample Type
Cells

Contents & Storage

Store Stop Solution, DNase I and 20X RT Enzyme Mix at -20°C. Store Lysis Solution, 2X SYBR™ RT Buffer, Fast SYBR™ Green PCR Master Mix at 4°C. Store Nuclease-free Water at room temperature.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text