Informational:Inquire about OEM or Commercial Supply version of this product here.
Invitrogen™

Cells-to-CT™ Stop Solution

Catalog number: 4402960
Have Questions?
Invitrogen™

Cells-to-CT™ Stop Solution

Catalog number: 4402960
Have Questions?
Catalog Number
4402960
Unit Size
1 mL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
44029601 mL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
This is a component of the Cells-to-CT™ Kits. One tube containing 1 mL is provided.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Form
Liquid
Green Features
Fewer resources used and less waste, Less hazardous
Product Line
Ambion™, Cells-to-CT™
Product Type
Stop Buffer
Shipping Condition
Dry Ice

Contents & Storage

Store Cells-to-CT™ Stop Solution at -20°C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text